Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ Stainless Steel Class 7 Individual Weights

Economical and designed for educational use.
Supplier: Troemner™ 80857695
Description
Adjusted to tolerance specified by ANSI/ASTM Class 7.
- Fractional masses of 200mg and over are made of stainless steel
- Fractional masses below 200mg are made of aluminum
Specifications
Individual Metric Economical Weight | |
Stainless Steel | |
7.85g/cm3 at 20°C | |
Cylindrical | |
1 |
ASTM Class 7 | |
10 g | |
10 g | |
Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction