Learn More
Troemner™ Stainless Steel Class 7 Individual Weights
SCI_ED
Economical and designed for educational use.
Supplier:  Troemner™ 80857700
 View more versions of this product
Catalog No. S40252A
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Adjusted to tolerance specified by ANSI/ASTM Class 7.
  • Fractional masses of 200mg and over are made of stainless steel
  • Fractional masses below 200mg are made of aluminum
Specifications
Specifications
Individual Metric Economical Sheet Weight with One Side Turned Up
Stainless Steel
7.85g/cm3 at 20°C
Flat
1
ASTM Class 7
200 mg
200 mg
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.