Learn More
AdipoGen Trolox, 53188-07-1, MFCD00006846, 500 mg
SDP
Supplier:  AdipoGen AGCR13639M500
Encompass_Preferred
MF: C14H18O4, MW: 250.3, ≥98%, Appearance: White to off-white or pale yellow solid. Cell permeable and water soluble derivative of vitamin E. Potent antioxidant. Efficiently prevents formation of different reactive oxygen species, such as singlet oxygen (1O2), superoxide anion (O2-) or hydrogen peroxide (H2O2). Commonly used as a standard or positive control in different antioxidant assays, such as ABTS decolorization and FRAP assays, measuring Trolox equivalent antioxidant capacity (TEAC). TEAC assays are used to measure antioxidant capacity of foods, beverages and supplements. Used in biological or biochemical applications to reduce oxidative stress or damage, such as inhibiting peroxynitrite mediated tyrosine nitration and mitochondrial proton permeability. Cytoprotective with low cytotoxicity for different cell lines. Shown to have anticancer activity and to be effective as adjunctive agent in cancer treatments.
Catalog No. 50-117-6131
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.