Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trovafloxacin mesylate, Tocris Bioscience™

Catalog No. p-4817484 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antibiotic; inhibits bacterial DNA synthesis

Fluoroquinolone antibiotic. Inhibits bacterial DNA topoisomerase IV and DNA gyrase and forms a stable quinolone-DNA complex with these enzymes which reversibly inhibits DNA synthesis. Displays potent activity against gram-positive and gram-negative bacteria. Increases the production of mitochondrial NO in immortalized hepatocytes; also increases mitochondrial Ca2+. Inhibits Panx-1 (IC50 ∽ 4μM).

Chemical Identifiers

CAS 147059-75-4
Molecular Formula C21H19F3N4O6S
Molecular Weight (g/mol) 512.46
InChI Key DYNZICQDCVYXFW-GIPYJWDTSA-N
Synonym trovafloxacin mesylate, trovan, trovafloxacin monomethanesulfonate, trovafloxacin methanesulfonate, unii-0p1lko80wn, 0p1lko80wn, trovafloxacin mesylate usan, trovan tn, trovafloxacin mesilate, dsstox_cid_25701
PubChem CID 62960
IUPAC Name 7-[(1S,5R)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid
SMILES CS(=O)(=O)O.C1C2C(C2N)CN1C3=C(C=C4C(=O)C(=CN(C4=N3)C5=C(C=C(C=C5)F)F)C(=O)O)F

Specifications

Quantity 10 mg
Formula Weight 512.46
Percent Purity >99%
Chemical Name or Material Trovafloxacin mesylate
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.