Learn More
Selleck Chemical LLC Troxerutin 50mg 7085-55-4
Supplier:  Selleck Chemical LLC S236950MG
Encompass_Preferred
Troxerutin, a natural bioflavonoid isolated from Sophora japonica, has been reported to have many benefits and medicinal properties. Troxerutin, also known as vitamin P4, is a tri-hydroxyethylated derivative of natural bioflavonoid rutins which can inhibit the production of reactive oxygen species (ROS) and depress ER stress-mediated NOD activation.
Catalog No. 50-136-2328
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.