Learn More
Selleck Chemical LLC Tryptamine-25mg
Supplier:  Selleck Chemical LLC S362725MG
Encompass_Preferred
Tryptamine is a monoamine alkaloid and may play a role as a neuromodulator or neurotransmitter. It is an agonist of hTAAR1. It acts as a non-selective serotonin receptor agonist and serotonin-norepinephrine-dopamine releasing agent (SNDRA).
Catalog No. 50-238-9993
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.