Learn More
Tosoh Bioscience LLC TSKGEL DEAE-3SW 7.5X75
Supplier: Tosoh Bioscience LLC 0007163
Column, Ion exchange chromatography; Tosoh; TSKgel DEAE-3SW; Matrix: hydrophilic silica; Stainless-steel; Counter ion: Clsup- Particle: 10u m; Pore size: 250angstrom pH: 2.0 -7.5; 7.5mm I.D. x 7.5cm length
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.