Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 4.6 X 150 MM
Supplier:  Tosoh Bioscience LLC 0021829
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 3 micrometer; Pore Size: 100 angstrom;4.6mm x 15cm Length; Stainless Steel
Catalog No. NC9618944
missing translation for 'QuantityLeft'