Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Protein C4-300 Guard Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7214154 Shop All MilliporeSigma Supelco Products
Click to view available options
Diameter (Metric) Inner:
2 mm
3.2 mm

Addition to the extensive line of Reversed Phase columns from Tosoh Bioscience

  • The high efficiency columns are designed for the optimal recovery and resolution of proteins such as recombinant proteins, antibody fragments or PEGylated proteins.
  • The 300 Å pore size of the TSKgel Protein C4-300 columns are ideal for the separation of proteins.
  • A particle size of 3 μm and optimized ligand density and alkyl length result in better protein and peptide resolution compared to other leading RP-C4 HPLC phases.
  • Latest surface modification techniques and endcapping of residual silanol groups reduce undesirable secondary interactions and peak tailing.
  • Shipped in acetonitrile.
  • 3% Carbon, endcapped with trimethylsilane
  • No limitation salt concentration
  • 0-100% organic solvent

Specifications

pH 1.5 - 7.5
Particle Size 3 μm
Surface Area 100 m2/g
Diameter (Metric) Inner 2 mm
Flow Rate 1 mL/min
For Use With (Equipment) L26 Per USP
Stationary Phase C4 (butyl)
Temperature (Metric) Maximum 10°C to 50°C
Material Stainless steel
Technique Reversed phase
Matrix Spherical silica
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.