Learn More
Tosoh Bioscience LLC TSKGEL SCX 6X150 5UM
Supplier: Tosoh Bioscience LLC 0007156
Column, Ion exchange chromatography; Tosoh; TSKgel SCX; Matrix: PS-DVB; Stainless-steel; Counter ion: Nasup+ Size: 5u m; (particle), 60angstrom (pore); Functional group: sulfonic acid; 6mm I.D. x 15cm length
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.