Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSNARE1 Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 240841AP150UL

Description
TSNARE1 Polyclonal antibody specifically detects TSNARE1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TSNARE1 | |
Polyclonal | |
Unconjugated | |
Q96NA8 | |
Rabbit | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TSNARE1 | |
Antigen affinity chromatography | |
RUO | |
203062 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction