Learn More
Thermo Scientific™ TSX Series High-Performance Undercounter Lab Refrigerators
For a better environment inside and out - Introducing our new undercounter lab refrigerator.
Supplier: Thermo Scientific™ TSX505GD
Description
Designed for your environment, new Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other research and medical products. Powered by V-drive, the TSX Series delivers low energy, decreased noise levels and improved performance and user experience. These new undercounter refrigerators are quiet and feature an improved user interface, automated locking and increased storage capacity and reliability.
Designed to maintain temperature uniformity to protect medical, pharmaceutical-grade products, laboratory media, reagents and more.- Unique V-drive technology is designed to save energy without compromising the protection of stored products
- U.S. Environmental Protection Agency SNAP (Significant New Alternatives Policy) compliant refrigerants
- Quiet operation at just 35dbA (43 during pull down)
- Reduced energy consumption
- Sleek exterior design and touch screen
- Bright, easy-to-adjust interior LED lighting
- Touchscreen controller with password protection
- Datalogging with USB download functionality
- Battery backup
- Choice of reversible glass or solid door
- Air or glycol temperature display
- Automated locking
- 1" rear access port
- Remote alarm contacts
- Audible and visual alarms: power fail, high temp, low temp, low battery, memory full, door ajar
- ADA compliant
- Compatible with most major electronic locks*
*Data on file.
Specifications
Alarm | |
45 Boxes per Level (9 x 5 Pattern), 225 Boxes per Shelf (5 Levels per Shelf, 2 Shelves per Unit), +45 Boxes (1 Additional Level Above Top Shelf) | |
1 | |
Energy efficient | |
TSX Series High-Performance Undercounter Refrigerator, Glass Door | |
CE, REACH, RoHS, UL | |
156 L | |
100/240 V | |
10 A Max / 3 A Steady State | |
26 x 23.6 x 31.8 in. (66 x 60 x 81 cm) | |
119 lb. | |
155 lb. | |
Full color TFT-LCD; 4.3”; 480 x 272 | |
Smart-Vue Compatible | |
52 cm | |
23 in. | |
58.4 cm | |
49.5 cm | |
23.6 in. | |
26 in. | |
66 cm |
Undercounter | |
3 | |
Single Glass | |
TSX Series | |
3°C to 7°C | |
5.5 cu. ft. | |
100 - 240 V 50-60 Hz | |
50/60 Hz | |
NEMA 6-15 P | |
20.5 x 19.5 x 23 in. (52 x 49.5 x 58.4 cm) | |
53.98 kg | |
71 kg | |
R744 | |
20.5 in. | |
31.8 in. | |
79.5 cm | |
19.5 in. | |
Automatic | |
60 cm | |
High-Performance Refrigerator |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.