Learn More
Saint Gobain Performance Plastics TUB NORPRENE 1/4 x 1/2 50FT/PK
Supplier:  Saint Gobain Performance Plastics AFL00019
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Saint-Gobain Norprene Industrial Tubing Formula A-60-G Diameter Inner 1/4 in. Thickness 0.125 in. Diameter Outer 1/2 in. Overall Length 50 ft. Maximum Pressure 19 psi
Catalog No. 50-206-9007