Learn More
Wilmad Labglass TUBE GAS COLL PTFE S/C 50ML
Supplier:  Wilmad Labglass LG8601T102
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Gas Collecting Tube, with PTFE Plugs. Capacity: 50mL; Body OD: 28mm; Body Length: 250mm; Stopcock Size: 2mm. Unit has a side outlet in body For a self sealing syringe stopper. Useful in gas chromatography or mass spectroscopy.
Catalog No. 50-806-326