Learn More
MilliporeSigma™ Supelco™ Tube Holders
For use with PAS-500 Micro Air Sampler in Air Monitoring Applications
Supplier:  MilliporeSigma™ Supelco™ 24868
 View more versions of this product
Catalog No. 11-101-1628
Explore all special offers
Add to Cart
Description
Description
Specifications
Specifications
8 mm Sorbent Tubes
Tube Holder
Air Monitoring
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.