Learn More
Ace Glass, Inc. Tube, filter, porosity D (10-20micron), (2)glass stopcock sidearms, 14/20 inner joints, No-Air
Supplier:  Ace Glass, Inc. 777418
Encompass_Preferred
Filter tube for NO-AIR™ applications, with two inner joints. Two, upper and lower side arms with either 2mm stopcocks or a 0-3 inlet valve. Two porosities are standard for the inner fritted disc for stronger and more precise flow.
Catalog No. 50-975-400
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.