Learn More
Perkin Elmer US LLC TUBG KIT-P40 REG 2ML/MIN PK12
Supplier:  Perkin Elmer US LLC N0680375
Encompass_Preferred
PVC Peristaltic Pump Tubing Kit, Red/Red i.d. 1.14mm 0.045in for 2mL/min. flow rate, Pkg. 12. Inner Diameter 0.045 in. (1.14 mm). Used to pump out the waste solution from the spray chamber. Kit includes PTFE TUBING.031 X.016 PER 1FT/0.3M.
Catalog No. 50-905-2652
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.