Learn More
Enterprise Technology Solutions Dynarex Oxygen Tubing 25 per case
SDP
Supplier:  Enterprise Technology Solutions DRX5101
Encompass_Preferred
The Dynarex Oxygen Tubing is available in two different styles, the Standard Lumen and the No Crush Lumen. The Oxygen Tubing with Standard Lumen has a smooth internal bore and measures 7-feet in length. The Oxygen Tubing with No Crush Lumen features a star-shaped interior cross section, reducing the likelihood of interrupting the flow of oxygen and measures 25-feet in length. The Standard Lumen comes with 50 per case, while the No Crush Lumen has 25 per case.
Catalog No. 50-240-7089
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.