Learn More
TARGETMOL CHEMICALS INC TUBULOSIDE A 10MG
Supplier:  TARGETMOL CHEMICALS INC T5S0281
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubuloside A ( 8.6 microM) can inhibit D-galactosamine-induced death of hepatocytes. Tubuloside A has NO radical-scavenging activity which possibly contributes to its anti-inflammatory effects.Tubuloside A shows stronger free radical scavenging activities than alpha-tocopherol on 11-diphenyl-2-picrylhydrazyl (DPPH) radical and xanthine/xanthine oxidase (XOD) generated superoxide anion radical (O2-.). purity: 99%
Catalog No. 50-257-6649
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.