Learn More
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals
Description
Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Brown to Yellow |
Flash Point | 17°C (63°F) |
Odor | Alcohol-like |
Linear Formula | W(OC2H5)6 |
Quantity | 25 mL |
UN Number | UN1170 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethyl acetate,hydrocarbons and esters. |
Formula Weight | 454.21 |
Percent Purity | 99.8% |
Show More |
Safety and Handling
Hazard Category | H225-H319-H335 |
Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
For Manufacturing and Laboratory Use. Not for beverage use
Your input is important to us. Please complete this form to provide feedback related to the content on this product.