Learn More
Perkin Elmer US LLC TurboMass Inert Gas Vent Kit
Supplier:  Perkin Elmer US LLC N6470045
Encompass_Preferred
The Mass Spectrometer Vent Kit eliminates the introduction of oxygen, water vapor, and other airborne contaminants which can degrade sensitivity and performance of the mass spectrometer. The Clarus GCs should be vented with UHP nitrogen (99.9995%). Helium should not be used as Helium molecules penetrate the glass sleeve on the photomultiplier tube, compromising the vacuum and lowering sensitivity.
Catalog No. 50-210-3977
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.