Learn More
Saint Gobain Performance Plastics Tube Tygon 1/8 x 3/16 50FT-15M/PK
Saint-GobainTygon S3 E3603 Flexible Tubings I.D. 1/8 in. O.D. 3/16 in. Wall thickness 1/32 in. Diameter Inner 1/8 in. Diameter Outer 3/16 in. Thickness 1/32 in. Overall Length 50 ft. 15 m Clear