Learn More
DuPont™ Tyvek™ 400 Disposable Lab Coats, USMCA Sourced
SDP
Catalog No. p-7138998 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Limited-use lab coats provide protection and durability. Meets USMCA standards.
DuPont™ Tyvek™ 400 provides lightweight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash
  • Abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance, operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
NAFTA
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Closure Type Snaps
Disposable Limited-use
Sterility Non-sterile
No. of Pockets 2
Model TY212SWH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.