Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U 73343, Tocris Bioscience™
GSA_VA

Catalog No. p-4817130 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Analog of U 73122 (Cat. No. 1268)

Analog of U 73122 ; can be used as a negative control. Inhibits Panx1 currents in HEK cells. Also inhibits vasopressin- and GTPγS-induced Ca2+ influx in hepatocytes.

Active Analog also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 142878-12-4
Molecular Formula C29H42N2O3
Molecular Weight (g/mol) 466.67
MDL Number MFCD00211221
InChI Key CJHWFIUASFBCKN-ZRJUGLEFSA-N
Synonym unii-s2c4j8704c, 1-6-17beta-3-methoxyestra-1,3,5 10-triene-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl-amino hexyl-2,5-pyrrolidine-dione, 1/6/2017, a-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17?-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl-2,5-pyrrolidine-dione, 1-6-8r,9s,13s,14s,17s-3-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta a phenanthren-17-yl amino hexyl pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-6-17beta-3-methoxyestra-1,3,5 10-trien-17-yl amino hexyl
PubChem CID 114825
IUPAC Name 1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]amino}hexyl)pyrrolidine-2,5-dione
SMILES COC1=CC=C2[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CCC2=C1)NCCCCCCN1C(=O)CCC1=O

Specifications

Quantity 10 mg
Formula Weight 466.66
Percent Purity >98%
Chemical Name or Material U 73343
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.