Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UK 383367, Tocris Bioscience™

Catalog No. p-4816505 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective BMP-1 (PCP) inhibitor

Potent and selective inhibitor of BMP-1 (procollagen C-proteinase; PCP) (IC50 = 44 nM). Selective for BMP-1 over MMPs 1, 2, 3, 9 and 14 (IC50 values are >10,000 nM for listed MMPs).

Chemical Identifiers

CAS 348622-88-8
Molecular Formula C15H24N4O4
Molecular Weight (g/mol) 324.381
InChI Key ARJCBSRIPGJMAD-LLVKDONJSA-N
PubChem CID 9818682
IUPAC Name 5-[(3R)-6-cyclohexyl-1-(hydroxyamino)-1-oxohexan-3-yl]-1,2,4-oxadiazole-3-carboxamide
SMILES C1CCC(CC1)CCCC(CC(=O)NO)C2=NC(=NO2)C(=O)N

Specifications

Quantity 1 mg
Synonym 5-1r-4-cyclohexyl-1-2-hydroxyamino-2-oxoethyl butyl-1,2,4-oxadiazole-3-carboxamide, 5-2r-5-cyclohexyl-1-hydroxycarbamoyl pentan-2-yl-1,2,4-oxadiazole-3-carboxamide, r-5-6-cyclohexyl-1-hydroxyamino-1-oxohexan-3-yl-1,2,4-oxadiazole-3-carboxamide, d0p2ts, uk hplc, 3r-6-cyclohexyl-n-hydroxy-3-1,2,4-oxadiazol-5-yl hexanamide, 3-aminocarbonyl-?-3-cyclohexylpropyl-n-hydroxy-1,2,4-oxadiazole-5-propanamide, 5-1r-4-cyclohexyl-1-2-hydroxyamino-2-oxoethyl butyl-1,2,4oxadiazole-3-carboxamide, 5-1 r-4-cyclohexyl-1-2-hydroxyamino-2-oxoethyl butyl-1,2,4-oxadiazole-3-carboxamide
Formula Weight 324.38
Percent Purity >99%
Chemical Name or Material UK 383367
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.