Learn More
TARGETMOL CHEMICALS INC UK-390957 5MG
Supplier:  TARGETMOL CHEMICALS INC T34994
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Uk-390957 is a sulfonamide compound with a high BPR due to binding to carbonic anhydrase. purity: 98%
Catalog No. 50-258-0240
missing translation for 'QuantityLeft'