Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UK 5099, Tocris Bioscience™

Catalog No. p-4816916 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MCT inhibitor; also inhibits mitochondrial pyruvate carrier

Inhibitor of plasma membrane monocarboxylate transporters (MCTs) and the mitochondrial pyruvate carrier (MPC).

Chemical Identifiers

CAS 56396-35-1
Molecular Formula C18H11N2O2-
Molecular Weight (g/mol) 287.298
InChI Key BIZNHCWFGNKBBZ-JLHYYAGUSA-M
Synonym 2-cyano-3-1-phenyl-1h-indol-3-yl-2-propenoic acid, 2e-2-cyano-3-1-phenylindol-3-yl prop-2-enoate
PubChem CID 71772548
IUPAC Name (E)-2-cyano-3-(1-phenylindol-3-yl)prop-2-enoate
SMILES C1=CC=C(C=C1)N2C=C(C3=CC=CC=C32)C=C(C#N)C(=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 288.3
Percent Purity >99%
Chemical Name or Material UK 5099
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.