Learn More
Perkin Elmer US LLC Ultra Clean High Capacity Triple Filter
Supplier:  Perkin Elmer US LLC N9306819
Encompass_Preferred
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. The triple trap is ideal for purifying carrier gas. It contains oxygen, moisture and hydrocarbon scrubbers in one easy to change economical cartridge. H2O Capacity: 1.8 g O2 Capacity: 75 mL HC Capacity: 4 g (as n-butane) Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: Inert carrier gas, He, H2, N2, Ar Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg
Catalog No. 50-210-5212
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.