Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ultra Pure Water Distributors

Use Thermo Scientific™ Ultra Pure Water Distributors with all SDI systems.

Supplier:  Thermo Scientific™ 50155458

 View more versions of this product

Catalog No. 13-998-284


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

Use Thermo Scientific™ Ultra Pure Water Distributors with all SDI systems.

  • Plastic
  • Connectors allow connection of shut-off taps
Specifications

Specifications

280 mm
50 mm
Distribution manifold: 1 inlet, 3 outlets
SDI Systems
170 mm
4 x R ¾ in. male thread (one inlet, three outlets)
PVC
Ultra Pure Water Distribution Manifold
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.