Learn More
Electron Microscopy Sciences Ultra Thin Carbon Tabs, 12 mm
Core material which is nonconductive cloth (70 um) with carbon Filler Conductive Adhesive (2 x 45μ). Total thickness is 160 µn;.
Diameter: 12 mm