Learn More
Electron Microscopy Sciences Quantifoil Holey Gold Film R 0.6/1 on Gold 300 mesh Grid
Supplier:  Electron Microscopy Sciences Q350AR1A
Encompass_Preferred
Newly developed ultrastable gold supports for electron cryomicroscopy will reduce the movement of frozen specimens during imaging. This improves image contrast and quality leading to better 3D reconstructions with less data.
During imaging at cryo-temperatures, traditional carbon supports move, particularly at the beginning of irradiation. This movement blurs images and makes it difficult to determine the structures of small and challenging molecules.
UltrAuFoilswere designed at MRC's Laboratory of Molecular Biology by Dr. Christopher J. Russo and Dr. Lori A. Passmore.
Catalog No. 50-192-7717
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.