Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecafluoroiodocyclohexane (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. p-7137865 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 355-69-1
Molecular Formula C6F11I
Molecular Weight (g/mol) 407.953
MDL Number MFCD00156095
InChI Key WYRAAMGCHCLHRD-UHFFFAOYSA-N
Synonym Perfluoroiodocyclohexane
PubChem CID 2759367
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-iodocyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)I)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 112°C
Quantity 1 g
Formula Weight 407.95
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecafluoroiodocyclohexane (stabilized with Copper chip)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.