Learn More
Glass Expansion UniFit Right Angle Connector (PKT 3)
Supplier: Glass Expansion GAZ0490
UniFit Right Angle Connector (PKT 3). Replacement set of 3 GazFit/UniFit right angle connectors for 4mm OD spray chamber drain port with capillary. Typically used with Glass Expansion’s IsoMist.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.