Learn More
Glass Expansion UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing (PKT. 10)
Supplier:  Glass Expansion NFT025
Encompass_Preferred
UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing sample tube. Package of 10 replacement sample lines. Compatible with Glass Expansion U-Series and DC Series Nebulizers. Used to carry sample from peristaltic pump tubing or mixing T to nebulizer. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For the same UniFit Connectors with the same ID but a longer sample tube length of 1500mm, NFTS-075. For UniFit Connector with 0.5mm ID, 1.3mm OD x 700mm long sample tube, NC1625341. For UniFit Connector with 0.18mm ID, 1.3mm OD x 700mm long sample tube, NC1973499.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2960
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.