Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Embedding Blocks
SDPSCI_ED

Embedding Blocks used for celloidin or paraffin sections.

Supplier:  United Scientific™ EB0001

 View more versions of this product

Catalog No. S90345A


Explore all special offers
Add to Cart

Description

Description

  • These embedding blocks are made of phenolic resin.
  • Top surface has 0.031 in. deep grid to provide a firm hold for embedded specimen.
  • Blocks are 0.75 in. high.
Specifications

Specifications

Embedding Matrix
0.5 x 0.5 in.
10 Pk.
Water-soluble Glycols and Resins
Used for Celloidin or Paraffin sections
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.