Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Litmus Paper
SDPSCI_ED

Available as Litmus paper and Litmus Granular in bottles.

Supplier:  United Scientific Supplies LGB100

 View more versions of this product

Catalog No. S99405


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • 100 strips each in matching color plastic vials
  • Individual strips are approximately 2 in. long and 0.25 in. wide

Specifications

Specifications

Granular
100 g Bottle
0.22 lb.
Measuring pH or acidity in liquids
Litmus Granular
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.