Learn More
Perkin Elmer US LLC Universal ATR Shoe - 6 mm
Supplier:  Perkin Elmer US LLC L1202050
Encompass_Preferred
PerkinElmer's Universal ATRs are excellent, easy-to-use accessories for the analysis of solids and liquids. These instruments produce high quality spectra through the use of a pressure arm, ensuring sample to sample and operator to operator reproducibility.
Catalog No. 50-211-1752
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.