Learn More
Restek™ Universal Press-Tight™ Connectors: Universal Y, Angled
SDP
Connect analytical columns, guard columns, transfer lines or detectors
$114.00 - $493.00
Products 4
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
06-717-753
SDP
View Documents
Restek™
20404
Universal Angled Y Press-Tight Connector
Pack of 3 for $493.00
Add to Cart
06-611-158
SDP
View Documents
Restek™
20403
Connector
Each for $114.00
Add to Cart
06-611-159
SDP
View Documents
Restek™
20403261
Deactivated Connector
Each for $129.50
Add to Cart
06-611-162
SDP
View Documents
Restek™
20487
Deactivated Connector
Each for $145.00
Add to Cart
Description
Description
  • Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
  • Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
  • Angle approximates the curvature of a capillary column, reducing the strain on column-end connections