Learn More
US Biomax Inc UNSTRAINED/MULTI ORGAN CANCER
SDP
Supplier:  US Biomax Inc BC000119B
Encompass
UNSTRAINED/Multiple organ cancer tissue arrayUNSTRAINED/Multiple organ cancer tissue arrayMultiple organ cancer tissue array, including pathology grade, TNM, clinical stage, Gleason grade and Gleason score, 190 cases/190 cores, replacing BC000119a, unstained slide
Catalog No. NC2301783
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.