Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, MP Biomedicals™

Catalog No. MP210320901 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1kg

Formed by the enzymatic reactions of the Kreb's cycle. Urea, MP Biomedicals is a principal protein metabolite end product of nitrogen metabolism and the major product for the removal of free ammonia (NH4+) in vivo. It is a mild agent usually used in the s

Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. It is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing. Isocyanic acid also reacts with side chains of lysine and arginine residues resulting in a protein that is unsuitable for many enzymatic digests. In addition, carbamylation often leads to confusing results from peptides having unexpected retention times and masses.

Urea is used in cell or tissue culture media to increase the osmolality.

Urea has also been used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Absorbance ≤0.2 (OD260nm and OD280 [7M aq soln])
Density 1.335g/cm3 at 20°C (Lit.)
pH 7.5 to 9.5 (7M aq. soln.)
Melting Point 130°C to 140°C
Boiling Point 135°C (literature) (decomposition)
Quantity 1kg
Solubility Information One gram dissolves in 1mL water, 10mL 95% ethanol (100mg/mL), 1mL boiling 95% ethanol, 20mL absolute ethanol (50mg/mL), 6mL methanol (166mg/mL), 2mL glycerol (500mg/mL); Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
Formula Weight 60.06
Physical Form White Prills
Grade Reagent
Vapor Pressure 0.000016hPa at 25°C (Lit.)
Show More Show Less
Recommended Storage Store at Room Temperature (15–30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.