Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea/Phosphoric/Dichromate Solution, Reagents

Urea/Phosphoric/Dichromate Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CU0005001A

Catalog No. 01-333-408


missing translation for 'QuantityLeft'
Add to Cart

Description

Description

Specifications

Specifications

Urea/Phosphoric/Dichromate Solution
7732-18-5, 7664-38-2, 57-13-6
Reagent
1.1971
1 L
Liquid
Clear Orange
100°C
Poly Bottle
<2
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.