Learn More
Innovative Research Inc US Bgle Sngl Pla Li Hep Vial
Supplier:  Innovative Research Inc IGCNBGPLASLIHVIAL
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Innovative Grade US Origin Canine Beagle Single Donor Plasma Li Heparin Vial - Innovative Grade US Origin Canine Beagle Plasma from Innovative Research is manufactured using whole blood collections from Beagles in USDA-approved facilities. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production.
Catalog No. 50-274-4542
missing translation for 'QuantityLeft'