Learn More
Innovative Research Inc US GnPig Comp Srm 10ml
Supplier:  Innovative Research Inc IGGPCSER10ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Innovative Grade US Origin Guinea Pig Complement Serum 10ml - Innovative Grade US Origin Guinea Pig Complement Serum from Innovative Research is manufactured using whole blood collections from guinea pigs. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity
Catalog No. 50-274-4529