Learn More
Innovative Research Inc US Mnky Cyno Comp Srm 50ml
Supplier: Innovative Research Inc IGMNCYCSER50ML
Innovative Grade US Origin Monkey Cynomolgus Complement Serum 50ml - Innovative Grade US Origin Monkey Cynomolgus Complement Serum from Innovative Research is manufactured using whole blood collections from type Cynomolgus monkeys. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the serum. This product is processed using a proprietary process that preserves complement activity. We do not recommend thawing this product for aliquoting, as this can impact the complement activity.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.