Learn More
Innovative Research Inc US Mnky Cyno Pla Li Hep 50ml
Supplier: Innovative Research Inc IGMNCYPLALIH50ML
Innovative Grade US Origin Monkey Cynomolgus Plasma Li Heparin 50ml - Innovative Grade US Origin Monkey Cynomolgus Plasma from Innovative Research is manufactured using whole blood collections from Cyno Monkeys. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.