Learn More
Innovative Research Inc US Ms B-C Sngl Whl Bld Na Hep
Supplier: Innovative Research Inc IGMSBCWBSNAHVIAL
Innovative Grade US Origin Mouse BALBC Single Donor Whole Blood Na Heparin Vial - Innovative Grade US Origin Mouse Balb C Single Donor Whole Blood from Innovative Research is collected per-order from type Mouse type Balb C. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.