Learn More
Innovative Research Inc US Pig Eye Whl W/ Skin Frsh 2
Supplier:  Innovative Research Inc IGPCEYFT1
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Innovative Grade US Origin Porcine Eye Whole With Skin Fresh Pair of 2 - Innovative Grade US Origin Porcine Eye Whole from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Porcine Eye, Whole is available in the following options Eye, Whole with Skin, Eye, Whole with Skin and Muscle, Eye, Whole with Skin and Optic Nerve attached. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4430
missing translation for 'QuantityLeft'