Learn More
Innovative Research Inc US Pig Vitreous Humor Fz 100ml
Supplier:  Innovative Research Inc IGPCVHZ100ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Innovative Grade US Origin Porcine Vitreous Humor Frozen 100ml - Innovative Grade US Origin Porcine Vitreous Humor from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4419
missing translation for 'QuantityLeft'