Learn More
Innovative Research Inc US Rat Sprg Dwly Pla K3E 50ml
Innovative Grade US Origin Rat Sprague Dawley Plasma K3 EDTA 50ml - Innovative Grade US Origin Rat Sprague Dawley Plasma from Innovative Research is manufactured using whole blood collections from Sprague Dawley rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.