Learn More
Innovative Research Inc US Rat Sprg Dwly Pla Li Hep
Supplier:  Innovative Research Inc IGRTSDPLALIH25ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Innovative Grade US Origin Rat Sprague Dawley Plasma Li Heparin 25ml - Innovative Grade US Origin Rat Sprague Dawley Plasma from Innovative Research is manufactured using whole blood collections from Sprague Dawley rats. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used for the production of the plasma.
Catalog No. 50-274-4402